
PSS-Octavinyl substituted

Synonym(s):
1,3,5,7,9,11,13,15-Octavinylpentacyclo[9.5.1.13,9.15,15.17,13]octasiloxane, Octavinyloctasilasesquioxane
Empirical Formula (Hill Notation):
C16H24O12Si8
CAS Number:
Molecular Weight:
633.04
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing