Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Proteomix
®
WAX-NP1.7 (1.7 µm) HPLC Columns
Products
Product Category
columns (5)
Manufacturer
pH Value
Product Line
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z777247
Proteomix
®
WAX-NP1.7 (1.7 µm) HPLC Columns,
L × I.D. 3 cm × 4.6 mm
Expand
View Pricing
Z777245
Proteomix
®
WAX-NP1.7 (1.7 µm) HPLC Columns,
L × I.D. 5 cm × 2.1 mm
Expand
View Pricing
Z777244
Proteomix
®
WAX-NP1.7 (1.7 µm) HPLC Columns,
L × I.D. 3 cm × 2.1 mm
Expand
View Pricing
Z777248
Proteomix
®
WAX-NP1.7 (1.7 µm) HPLC Columns,
L × I.D. 5 cm × 4.6 mm
Expand
View Pricing
Z777243
Proteomix
®
WAX-NP1.7 (1.7 µm) HPLC Columns,
L × I.D. 1 cm × 2 mm
Expand
View Pricing
Z777246
Proteomix
®
WAX-NP1.7 (1.7 µm) HPLC Columns,
L × I.D. 1 cm × 4 mm
Expand
View Pricing