![Proscillaridin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/138/522/ca93b2f3-bdf2-492b-8db2-1442a14f1361/640/ca93b2f3-bdf2-492b-8db2-1442a14f1361.png)
Proscillaridin A
![Proscillaridin A phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/138/522/ca93b2f3-bdf2-492b-8db2-1442a14f1361/640/ca93b2f3-bdf2-492b-8db2-1442a14f1361.png)
Synonym(s):
Proscillaridin A, 3-(6-Deoxy-α-L-mannopyranosyloxyl)-14-hydroxybufa-4,20,22-trienolide, Scillarenin 3β-rhamnoside
Empirical Formula (Hill Notation):
C30H42O8
CAS Number:
Molecular Weight:
530.65
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing