
Propylene carbonate

Synonym(s):
1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one, Propylene carbonate
Empirical Formula (Hill Notation):
C4H6O3
CAS Number:
Molecular Weight:
102.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing