Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium chromate
Synonym(s):
Potassium chromate
Linear Formula:
K
2
CrO
4
CAS Number:
7789-00-6
Molecular Weight:
194.19
EC Number:
232-140-5
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (3)
Supelco (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade
ACS reagent (1)
Puriss (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
216615
Potassium chromate,
ACS reagent, ≥99.0%
Expand
View Pricing
1.04952
Potassium chromate,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
12249
Potassium chromate,
puriss., ≥99%
Expand
View Pricing
529508
Potassium chromate,
≥99.95% trace metals basis
Expand
View Pricing