Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(vinylidene fluoride-
co
-hexafluoropropylene)
Synonym(s):
PVDF-HFP
Linear Formula:
(-CH
2
CF
2
-)
x
[-CF
2
CF(CF
3
)-]
y
CAS Number:
9011-17-0
Products
Physical Form
Polymer Composition
Polymer Type
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
427160
Poly(vinylidene fluoride-
co
-hexafluoropropylene),
average M
w
~400,000, average M
n
~130,000, pellets
Expand
View Pricing
427179
Poly(vinylidene fluoride-
co
-hexafluoropropylene),
average M
w
~455,000, average M
n
~110,000, pellets
Expand
View Pricing
427187
Poly(vinylidene fluoride-
co
-hexafluoropropylene),
pellets
Expand
View Pricing