
Polystyrene-block-polybutadiene-block-polystyrene

Linear Formula:
[CH2CH(C6H5)]x (CH2CH=CHCH2)y [CH2CH(C6H5)]x
CAS Number:
Products
Physical Form
Chemical Composition
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing