Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(acrylic acid sodium salt)
Synonym(s):
Poly(sodium acrylate), Sodium polyacrylate
CAS Number:
9003-04-7
Products
Brand
Sigma-Aldrich (2)
Supelco (1)
pH Value
Physical Form
Format
Quality Segment
Polymer Composition
Polymer Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
447013
Poly(acrylic acid sodium salt),
average M
w
~5,100 by GPC, powder
Expand
View Pricing
420344
Poly(acrylic acid sodium salt),
average M
w
~2,100
Expand
View Pricing
81123
Poly(acrylic acid sodium salt),
analytical standard, for GPC, 16,000
Expand
View Pricing