Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pipette tips, for Pipetman
®
P2/P10 and Eppendorf
®
Research/Reference pipettors, ultramicro, extra long
Products
Product Category
pipette tips (3)
Manufacturer
Color
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
AXYTXL10R
Pipette tips, for Pipetman
®
P2/P10 and Eppendorf
®
Research/Reference pipettors, ultramicro, extra long,
size 0.1-10 μL, pkg of 5x960tips/cs, racked, clear
Expand
View Pricing
AXYTXL10
Pipette tips, for Pipetman
®
P2/P10 and Eppendorf
®
Research/Reference pipettors, ultramicro, extra long,
size 0.1-10 μL, pkg of 20x1000tips/cs, clear
Expand
View Pricing
AXYTXL10RS
Pipette tips, for Pipetman
®
P2/P10 and Eppendorf
®
Research/Reference pipettors, ultramicro, extra long,
size 0.1-10 μL, pkg of 5x960tips/cs, racked, clear, sterile
Expand
View Pricing