Skip to Content
Merck
Phytic acid sodium salt hydrate ≥90% phosphorus (P) basis (dry basis)

Phytic acid sodium salt hydrate

Phytic acid sodium salt hydrate ≥90% phosphorus (P) basis (dry basis)
Synonym(s):
myo-Inositol hexakis(dihydrogen phosphate), Inositol hexakisphosphate, Inositol hexaphosphoric acid, InsP6
Empirical Formula (Hill Notation):
C6H18O24P6 · xNa+ · yH2O
CAS Number:
Molecular Weight:
660.04 (anhydrous free acid basis)
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Showing 1-2 of 2