Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pellicon
®
XL Cassette with Durapore
®
Membrane
Products
Product Category
filters (4)
Brand
Millipore (4)
pH Value
Length (cm)
Length (in)
Surface Area cm²
Quality Segment
Membrane Material
Pore size (µm)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
PXHVMPC50
Pellicon
®
XL Cassette with Durapore
®
Membrane,
filtration area 50 cm
2
, pore size 0.45 μm
Expand
View Pricing
PXGVPPC50
Pellicon
®
XL Cassette with Durapore
®
Membrane,
filtration area 50 cm
2
, pore size 0.22 μm
Expand
View Pricing
PXDVPPC50
Pellicon
®
XL Cassette with Durapore
®
Membrane,
filtration area 50 cm
2
, pore size 0.65 μm
Expand
View Pricing
PXVVPPC50
Pellicon
®
XL Cassette with Durapore
®
Membrane,
filtration area 50 cm
2
, pore size 0.1 μm
Expand
View Pricing