
Oxypeucedanin

Synonym(s):
4-[[(2S)-3,3-Dimethyl-2-oxiranyl]methoxy]-7H-furo[3,2-g][1]benzopyran-7-one
Empirical Formula (Hill Notation):
C16H14O5
CAS Number:
Molecular Weight:
286.28
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing