Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ohaus
®
Scout
®
Portable Touchscreen Balance
Products
Product Category
balances (12)
Manufacturer
Length (cm)
Weighing Capacity (g)
Resolution (g)
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
OH30253068
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX6201, weighing capacity 6200 g, resolution: 0.1 g, AC/DC input 230 V
Expand
View Pricing
OH30253063
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX1202, weighing capacity 1200 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
Expand
View Pricing
OH30253059
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX223, weighing capacity 220 g, resolution: 0.001 g, AC/DC input 230 V
Expand
View Pricing
OH30253058
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX123, weighing capacity 120 g, resolution: 0.001 g, AC/DC input 230 V
Expand
View Pricing
OH30253065
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX421, weighing capacity 420 g, resolution: 0.1 g, AC/DC input 230 V
Expand
View Pricing
OH30253060
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX222, weighing capacity 220 g, resolution: 0.01 g, AC/DC input 230 V
Expand
View Pricing
OH30253064
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX2202, weighing capacity 2200 g, resolution: 0.01 g, AC/DC input 230 V
Expand
View Pricing
OH30253066
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX621, weighing capacity 620 g, resolution: 0.1 g, AC/DC input 230 V
Expand
View Pricing
OH30253061
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX422, weighing capacity 420 g, resolution: 0.01 g, AC/DC input 230 V
Expand
View Pricing
OH30253062
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX622, weighing capacity 620 g, resolution: 0.01 g, AC/DC input 230 V
Expand
View Pricing
OH30253067
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX2201, weighing capacity 2200 g, resolution: 0.1 g, AC/DC input 230 V
Expand
View Pricing
OH30253069
Ohaus
®
Scout
®
Portable Touchscreen Balance,
Model STX8200, weighing capacity 8200 g, resolution: 1 g, AC/DC input 230 V
Expand
View Pricing