Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nunc
®
barcoded latch rack
Products
Product Category
racks (7)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
TMO331825-BR
Nunc
®
barcoded latch rack,
for 2.0 mL cryogenic tubes, plastic, 42 positions, case of 10 ea
Expand
View Pricing
TMO331827-BR
Nunc
®
barcoded latch rack,
for 1.0-2.0 mL cryogenic tubes, plastic, 42 positions, case of 10 ea
Expand
View Pricing
TMO331826-BR
Nunc
®
barcoded latch rack,
for 5.0 mL cryogenic tubes, plastic, 30 positions, case of 10 ea
Expand
View Pricing
TMO331828-BR
Nunc
®
barcoded latch rack,
for 3.6-5.0 mL cryogenic tubes, plastic, 30 positions, case of 10 ea
Expand
View Pricing
TMO331835-BR
Nunc
®
barcoded latch rack,
for 2.0 mL internal thread universal tubes, plastic, 42 positions, case of 10 ea
Expand
View Pricing
TMO331830-BR
Nunc
®
barcoded latch rack,
for 1.8 mL external thread universal tubes, plastic, 48 positions, case of 10 ea
Expand
View Pricing
TMO331840-BR
Nunc
®
barcoded latch rack,
for low profile 300 mL external thread universal tubes, plastic, 30 positions, case of 10 ea
Expand
View Pricing