
Norbuprenorphine-D3 solution

Empirical Formula (Hill Notation):
C25H32D3NO4
CAS Number:
Molecular Weight:
416.57
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique