Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
,
N
-Dimethyl-
p
-phenylenediamine dihydrochloride
Synonym(s):
4-Amino-
N
,
N
-dimethylaniline dihydrochloride, DMPD · 2HCl, DMPPDA · 2HCl
Linear Formula:
(CH
3
)
2
NC
6
H
4
NH
2
· 2HCl
CAS Number:
536-46-9
Molecular Weight:
209.12
EC Number:
208-635-7
Products
Product Category
stains (1)
substrates (1)
Brand
Millipore (1)
Sigma-Aldrich (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
D4139
N
,
N
-Dimethyl-
p
-phenylenediamine dihydrochloride,
≥99.0% purity (titration), powder
Expand
View Pricing
07770
N
,
N
-Dimethyl-
p
-phenylenediamine dihydrochloride,
suitable for microbiology, ≥99.0%
Expand
View Pricing