Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
N
,
N
-Dimethyl-4,4′-azodianiline
Synonym(s):
4-(4-Dimethylaminophenylazo)aniline, 4-Amino-4′-(dimethylamino)azobenzene, 4-Amino-4′-dimethylaminoazobenzene,
p
,
p
′-DMPA-aniline,
p
,
p
′-Dimapa-aniline, Disperse Black 3
Linear Formula:
(CH
3
)
2
NC
6
H
4
N=NC
6
H
4
NH
2
CAS Number:
539-17-3
Molecular Weight:
240.30
EC Number:
208-712-5
Products
Product Category
stains (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
379298
N
,
N
-Dimethyl-4,4′-azodianiline,
97%
Expand
View Pricing