Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
NMP
Synonym(s):
1-Methyl-2-pyrrolidone,
N
-Methyl-2-pyrrolidone, NMP, 1-Methyl-2-pyrrolidinone
Empirical Formula (Hill Notation):
C
5
H
9
NO
CAS Number:
872-50-4
Molecular Weight:
99.13
EC Number:
212-828-1
Products
Product Category
solvents (9)
Brand
Sigma-Aldrich (5)
Supelco (6)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
ACS reagent (1)
Anhydrous (1)
Biotechnology (1)
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
443778
NMP,
ACS reagent, ≥99.0%
Expand
View Pricing
M79603
NMP,
ReagentPlus
®
, 99%
Expand
View Pricing
270458
NMP,
suitable for HPLC, ≥99%
Expand
View Pricing
494496
NMP,
biotech. grade, ≥99.7%
Expand
View Pricing
1.02497
NMP,
for headspace gas chromatography SupraSolv
®
Expand
View Pricing
8.06072
NMP,
EMPLURA
®
Expand
View Pricing
5.89597
NMP,
99.5%, anhydrous
Expand
View Pricing
1.00574
NMP,
for Peptide Synthesis
Expand
View Pricing
5.43899
NMP,
for liquid chromatography LiChrosolv
®
Expand
View Pricing
PHR1352
NMP,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
78769
NMP,
analytical standard
Expand
View Pricing