Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nitromethane
Synonym(s):
Nitromethane
Empirical Formula (Hill Notation):
CH
3
NO
2
CAS Number:
75-52-5
Molecular Weight:
61.04
EC Number:
200-876-6
Products
Product Category
solvents (4)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Special Grade
ACS reagent (1)
Absolute (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
360554
Nitromethane,
ACS reagent, ≥95%
Expand
View Pricing
230731
Nitromethane,
ReagentPlus
®
, ≥99.0%
Expand
View Pricing
270423
Nitromethane,
suitable for HPLC, ≥96%
Expand
View Pricing
8.20894
Nitromethane,
for synthesis
Expand
View Pricing
73478
Nitromethane,
puriss., absolute, over molecular sieve, ≥98.5% (GC)
Expand
View Pricing