Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-20 of 39
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF92920923
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
foil, 100 x 100mm, 0.5mm thickness, annealed
Expand
View Pricing
GF56315459
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
foil, 50 x 50mm, 0.5mm thickness, annealed
Expand
View Pricing
GF32319176
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
5m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF29247419
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
foil, 100 x 100mm, 2.0mm thickness, annealed
Expand
View Pricing
GF66590275
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
200mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF86640240
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
50g lump, 10mm max. lump size, pellets
Expand
View Pricing
GF07374966
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
foil, 100 x 100mm, 1.0mm thickness, annealed
Expand
View Pricing
GF86115348
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
100mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF21870659
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
foil, 150 x 150mm, 1.0mm thickness, annealed
Expand
View Pricing
GF66817301
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
1000mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF36499449
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
1000mm rod, 3.2mm diameter, as drawn
Expand
View Pricing
GF87840197
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
1000mm tube, 15mm outside diameter, 0.4mm wall thickness, 14.2mm inside diameter, as drawn
Expand
View Pricing
GF90976809
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
500mm tube, 15mm outside diameter, 0.4mm wall thickness, 14.2mm inside diameter, as drawn
Expand
View Pricing
GF89872633
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
10m wire, 0.5mm diameter, as drawn
Expand
View Pricing
GF42202921
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
foil, 76 x 76mm, 3.5mm thickness, annealed
Expand
View Pricing
GF01658998
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
foil, 50 x 50mm, 1.0mm thickness, annealed
Expand
View Pricing
GF85155093
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
200mm rod, 25mm diameter, annealed
Expand
View Pricing
GF74109001
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
500mm rod, 5.0mm diameter, as drawn
Expand
View Pricing
GF12377684
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
100mm rod, 3.2mm diameter, as drawn
Expand
View Pricing
GF42956208
Nickel-iron-molybdenum alloy, Ni80Fe15.5Mo4.5,
200g lump, 10mm max. lump size, pellets
Expand
View Pricing
Page 1 of 2
Page 1 of 2