Skip to Content
Merck
Neopentyl chloroformate 97%

Neopentyl chloroformate

Neopentyl chloroformate 97%
Synonym(s):
2,2-Dimethylpropyl chloroformate
Linear Formula:
(CH3)3CCH2OCOCl
CAS Number:
Molecular Weight:
150.60

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing