Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Monel
®
alloy 400, Ni65Cu33Fe2
Products
Manufacturer
Physical Form
Diameter (mm)
ID (mm)
Length (m)
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF83915365
Monel
®
alloy 400, Ni65Cu33Fe2,
mesh, 100 x 100mm, 0.38mm nominal aperture, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh
Expand
View Pricing
GF89420834
Monel
®
alloy 400, Ni65Cu33Fe2,
mesh, 150 x 150mm, 0.38mm nominal aperture, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh
Expand
View Pricing
GF77604059
Monel
®
alloy 400, Ni65Cu33Fe2,
mesh, 300 x 300mm, 0.38mm nominal aperture, 0.25mm wire diameter, 40 x 40 wires/inch, 0.37 open area, plain weave mesh
Expand
View Pricing
GF54083268
Monel
®
alloy 400, Ni65Cu33Fe2,
500mm tube, 1.5mm outside diameter, 0.25mm wall thickness, 1mm inside diameter, as drawn
Expand
View Pricing
GF43843204
Monel
®
alloy 400, Ni65Cu33Fe2,
500mm tube, 0.6mm outside diameter, 0.1mm wall thickness, 0.4mm inside diameter, as drawn
Expand
View Pricing
GF04386809
Monel
®
alloy 400, Ni65Cu33Fe2,
100mm tube, 0.6mm outside diameter, 0.1mm wall thickness, 0.4mm inside diameter, as drawn
Expand
View Pricing
GF13996345
Monel
®
alloy 400, Ni65Cu33Fe2,
1000mm wire, 1.0mm diameter, annealed, straight wire
Expand
View Pricing
GF96750029
Monel
®
alloy 400, Ni65Cu33Fe2,
5m wire, 1.5mm diameter, as drawn
Expand
View Pricing
GF17953847
Monel
®
alloy 400, Ni65Cu33Fe2,
foil, 100 x 100mm, 0.2mm thickness, annealed
Expand
View Pricing
GF47240096
Monel
®
alloy 400, Ni65Cu33Fe2,
500mm tube, 0.8mm outside diameter, 0.1mm wall thickness, 0.6mm inside diameter, as drawn
Expand
View Pricing
GF30931376
Monel
®
alloy 400, Ni65Cu33Fe2,
25m wire, 0.125mm diameter, as drawn
Expand
View Pricing
GF87347201
Monel
®
alloy 400, Ni65Cu33Fe2,
foil, 1m coil, 0.075mm thickness, annealed
Expand
View Pricing
GF25338900
Monel
®
alloy 400, Ni65Cu33Fe2,
foil, 4mm disks, 0.030mm thickness, annealed
Expand
View Pricing
GF49790739
Monel
®
alloy 400, Ni65Cu33Fe2,
500mm tube, 6.35mm outside diameter, 1.5mm wall thickness, 3.35mm inside diameter, as drawn
Expand
View Pricing
GF05012086
Monel
®
alloy 400, Ni65Cu33Fe2,
1000mm tube, 0.8mm outside diameter, 0.1mm wall thickness, 0.6mm inside diameter, as drawn
Expand
View Pricing
GF13788377
Monel
®
alloy 400, Ni65Cu33Fe2,
200mm tube, 1.27mm outside diameter, 0.25mm wall thickness, 0.77mm inside diameter, as drawn
Expand
View Pricing
GF43365944
Monel
®
alloy 400, Ni65Cu33Fe2,
25m wire, 1.5mm diameter, as drawn
Expand
View Pricing
GF68423890
Monel
®
alloy 400, Ni65Cu33Fe2,
rod, 500mm x 12.7mm diameter, as drawn
Expand
View Pricing
GF83203185
Monel
®
alloy 400, Ni65Cu33Fe2,
100mm tube, 6.35mm outside diameter, 1.5mm wall thickness, 3.35mm inside diameter, as drawn
Expand
View Pricing
GF03718470
Monel
®
alloy 400, Ni65Cu33Fe2,
100mm tube, 2.5mm outside diameter, 0.18mm wall thickness, 2.14mm inside diameter, as drawn
Expand
View Pricing
Page 1 of 11
Page 1 of 11