Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler-Toledo SevenGo
™
Duo pH/conductivity SG23
Products
Manufacturer
pH Value
Length (m)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
MT51302610
Mettler-Toledo SevenGo
™
Duo pH/conductivity SG23,
SG68-Basic, no electrode
Expand
View Pricing
MT51302612
Mettler-Toledo SevenGo
™
Duo pH/conductivity SG23,
SG68 field kit, InLab
®
Expert Pro-ISM and InLab
®
605-ISM, cable L 2 m
Expand
View Pricing
MT51302613
Mettler-Toledo SevenGo
™
Duo pH/conductivity SG23,
SG68 field kit, InLab
®
Expert Pro-ISM and InLab
®
605-ISM, cable L 5 m
Expand
View Pricing
MT51302611
Mettler-Toledo SevenGo
™
Duo pH/conductivity SG23,
SG68 kit, InLab
®
Expert Pro-ISM and InLab
®
605-ISM
Expand
View Pricing