Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methyl linolelaidate
Synonym(s):
Linolelaidic acid methyl ester, Methyl
trans
,
trans
-9,12-octadecadienoate
Empirical Formula (Hill Notation):
C
19
H
34
O
2
CAS Number:
2566-97-4
Molecular Weight:
294.47
EC Number:
219-901-7
Products
Product Category
lipids (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Biological Source
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
L2251
Methyl linolelaidate,
Expand
View Pricing
62155
Methyl linolelaidate,
analytical standard
Expand
View Pricing