Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methylamine solution
Synonym(s):
Monomethylamine
Linear Formula:
CH
3
NH
2
CAS Number:
74-89-5
Molecular Weight:
31.06
Products
Product Category
organic bases (3)
Brand
Sigma-Aldrich (4)
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Special Grade
Absolute (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
395056
Methylamine solution,
2.0 M in THF
Expand
View Pricing
534102
Methylamine solution,
33 wt. % in absolute ethanol ((denatured with 1% toluene))
Expand
View Pricing
395048
Methylamine solution,
2.0 M in methanol
Expand
View Pricing
426466
Methylamine solution,
40 wt. % in H
2
O
Expand
View Pricing