Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methyl α-D-mannopyranoside
Synonym(s):
α-Methyl D-mannoside, Methyl-α-D-mannopyranoside
Empirical Formula (Hill Notation):
C
7
H
14
O
6
CAS Number:
617-04-9
Molecular Weight:
194.18
EC Number:
210-502-3
Products
Product Category
carbohydrates (2)
bioactive small molecules (1)
monosaccharides (1)
Brand
Millipore (1)
Sigma-Aldrich (2)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Application
Optical Isomer
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
M6882
Methyl α-D-mannopyranoside,
≥99.0% (HPLC)
Expand
View Pricing
462711
Methyl α-D-mannopyranoside,
Methyl-α-D-mannopyranoside, CAS 617-04-9, is a substrate for α1,2-Mannosyltransferase (Cat. No. 444125).
Expand
View Pricing
67770
Methyl α-D-mannopyranoside,
≥99.0%, suitable for microbiology, enables differentiation between species of
Listeria
Expand
View Pricing