![Methotrexate for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/315/821/a211b71d-f9cf-45ed-86c8-a0b657747574/640/a211b71d-f9cf-45ed-86c8-a0b657747574.png)
Methotrexate for system suitability
![Methotrexate for system suitability European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/315/821/a211b71d-f9cf-45ed-86c8-a0b657747574/640/a211b71d-f9cf-45ed-86c8-a0b657747574.png)
Synonym(s):
Methotrexate, (+)-Amethopterin, 4-Amino-10-methylfolic acid, 4-Amino-N10-methylpteroyl-L-glutamic acid, Antifolan, MTX, Methylaminopterin
Empirical Formula (Hill Notation):
C20H22N8O5
CAS Number:
Molecular Weight:
454.44
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing