![Losartan potassium British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/510/939/29fb0438-fda4-40c2-adf0-39bdd877abcb/640/29fb0438-fda4-40c2-adf0-39bdd877abcb.png)
Losartan potassium
![Losartan potassium British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/510/939/29fb0438-fda4-40c2-adf0-39bdd877abcb/640/29fb0438-fda4-40c2-adf0-39bdd877abcb.png)
Synonym(s):
2-Butyl-4-chloro-1-{[2′-(1H-tetrazol-5-yl)(1,1′-biphenyl)-4-yl]methyl}-1H-imidazole-5-methanol monopotassium salt, 2-butyl-4-chloro-1-[[2′-(2H-tetrazol-5-yl)[1,1′-biphenyl]-4-yl]methyl]- 1H-Imidazole-5-methanol potassium salt, DuP 753, MK 954, Potassium 5-(4′-((2-butyl-4-chloro-5-(hydroxymethyl)-1H-imidazol-1-yl)methyl)-[1,1′-biphenyl]-2-yl)tetrazol-1-ide, Losartan potassium
Empirical Formula (Hill Notation):
C22H22ClKN6O
CAS Number:
Molecular Weight:
461.00
Products
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing