L-Methionine p-nitroanilide
Empirical Formula (Hill Notation):
C11H15N3O3S
CAS Number:
Molecular Weight:
269.32
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Quality Segment
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing