Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lenz
®
round bottom flasks
Products
Product Category
flasks (12)
Manufacturer
Agency
Capacity (mL)
Closure Type
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
Z331406
Lenz
®
round bottom flasks,
capacity 25 mL, joint: ST/NS 14/23
Expand
View Pricing
Z331449
Lenz
®
round bottom flasks,
capacity 50 mL, joint: ST/NS 29/32
Expand
View Pricing
Z331465
Lenz
®
round bottom flasks,
capacity 250 mL, joint: ST/NS 29/32
Expand
View Pricing
Z331457
Lenz
®
round bottom flasks,
capacity 100 mL, joint: ST/NS 29/32
Expand
View Pricing
Z331481
Lenz
®
round bottom flasks,
capacity 1,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z331473
Lenz
®
round bottom flasks,
capacity 500 mL, joint: ST/NS 29/32
Expand
View Pricing
Z331422
Lenz
®
round bottom flasks,
capacity 100 mL, joint: ST/NS 14/23
Expand
View Pricing
Z331503
Lenz
®
round bottom flasks,
capacity 2,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z331511
Lenz
®
round bottom flasks,
capacity 4,000 mL, joint: ST/NS 29/32
Expand
View Pricing
Z331414
Lenz
®
round bottom flasks,
capacity 50 mL, joint: ST/NS 14/23
Expand
View Pricing
Z331392
Lenz
®
round bottom flasks,
joint: ST/NS 14/23, capacity 10 mL
Expand
View Pricing
Z331430
Lenz
®
round bottom flasks,
capacity 250 mL, joint: ST/NS 14/23
Expand
View Pricing