Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Canavanine sulfate salt
Synonym(s):
L-α-Amino-γ-(guanidinooxy)butyric acid sulfate salt
Empirical Formula (Hill Notation):
C
5
H
12
N
4
O
3
· H
2
SO
4
CAS Number:
2219-31-0
Molecular Weight:
274.25
EC Number:
218-728-4
Products
Product Category
peptides (1)
Brand
Sigma-Aldrich (1)
Biological Source
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Tissue
Quality Segment
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C9758
L-Canavanine sulfate salt,
≥99% (TLC), powder
Expand
View Pricing