Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Julabo
®
adaptors
Products
Manufacturer
Package Size
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z743289
Julabo
®
adaptors,
m16x1 male to bsp 1/2 inch female, pack of 1
Expand
View Pricing
Z743286
Julabo
®
adaptors,
m16x1 female to tube 1/2 inch male, pack of 2
Expand
View Pricing
Z743279
Julabo
®
adaptors,
m16x1 female to m16x1 female, pack of 2
Expand
View Pricing
Z743284
Julabo
®
adaptors,
m16x1 female to npt 1/4 inch male, pack of 2
Expand
View Pricing
Z743287
Julabo
®
adaptors,
m16x1 female to tube 1/4 inch male, pack of 2
Expand
View Pricing
Z743273
Julabo
®
adaptors,
barbed fittings, for 12 mm i.d. tubing, pack of 2
Expand
View Pricing
Z743278
Julabo
®
adaptors,
m16 x1 female to npt 3/8 inch male, pack of 2
Expand
View Pricing
Z743274
Julabo
®
adaptors,
barbed fittings, for 8 mm i.d. tubing, pack of 2
Expand
View Pricing
Z743282
Julabo
®
adaptors,
m16x1 female to npt 1/4 inch female, pack of 2
Expand
View Pricing
Z743292
Julabo
®
adaptors,
m16x1 male to npt 1/4 inch female, pack of 2
Expand
View Pricing
Z743296
Julabo
®
adaptors,
twin distributing, one m16x1 female to 2 x m16x1 male, pack of 1
Expand
View Pricing
Z743285
Julabo
®
adaptors,
m16x1 female to npt 3/8 inch female, pack of 2
Expand
View Pricing
Z743272
Julabo
®
adaptors,
barbed fittings, for 10 mm i.d. tubing, pack of 2
Expand
View Pricing
Z743276
Julabo
®
adaptors,
for metal tubing, m10 male to m16 male, pack of 1
Expand
View Pricing
Z743281
Julabo
®
adaptors,
m16x1 female to npt 1/2 inch male, pack of 2
Expand
View Pricing
Z743290
Julabo
®
adaptors,
m16x1 male to bsp 3/4 inch female, pack of 1
Expand
View Pricing
Z743293
Julabo
®
adaptors,
quad distributing, 1 1/4 inch female with barbed fittings, for 1 inch i.d. tubing, pack of 2
Expand
View Pricing
Z743280
Julabo
®
adaptors,
m16x1 female to npt 1/2 inch female, pack of 2
Expand
View Pricing
Z743295
Julabo
®
adaptors,
twin distributing with barbed fittings, for 8 mm i.d. tubing, pack of 1
Expand
View Pricing
Z743277
Julabo
®
adaptors,
for metal tubing, m16x1 male to m16x1 male, pack of 1
Expand
View Pricing
Page 1 of 2
Page 1 of 2