Skip to Content
Merck
Iron(II) bromide AnhydroBeads™, −10 mesh, 99.999% trace metals basis

Iron(II) bromide

Iron(II) bromide AnhydroBeads™, −10 mesh, 99.999% trace metals basis
Synonym(s):
Ferrous bromide
Linear Formula:
FeBr2
CAS Number:
Molecular Weight:
215.65
EC Number:

Products

Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reaction Type
Reagent Type
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing