Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Indium-tin alloy, In50Sn50
Products
Manufacturer
Physical Form
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
GF40761097
Indium-tin alloy, In50Sn50,
foil, 25 x 25mm, 0.1mm thickness
Expand
View Pricing
GF35925521
Indium-tin alloy, In50Sn50,
foil, 25 x 25mm, 0.05mm thickness, not light tested (nlt)
Expand
View Pricing
GF96978245
Indium-tin alloy, In50Sn50,
foil, 50 x 50mm, 0.25mm thickness
Expand
View Pricing
GF43244276
Indium-tin alloy, In50Sn50,
foil, 100 x 100mm, 0.25mm thickness
Expand
View Pricing
GF57926638
Indium-tin alloy, In50Sn50,
foil, 50 x 50mm, 0.05mm thickness, not light tested (nlt)
Expand
View Pricing
GF37626587
Indium-tin alloy, In50Sn50,
foil, 50 x 50mm, 0.1mm thickness
Expand
View Pricing
GF49453566
Indium-tin alloy, In50Sn50,
foil, 100 x 100mm, 0.05mm thickness, not light tested (nlt)
Expand
View Pricing
GF71607012
Indium-tin alloy, In50Sn50,
foil, 25 x 25mm, 0.25mm thickness
Expand
View Pricing
GF89928321
Indium-tin alloy, In50Sn50,
foil, 100 x 100mm, 0.1mm thickness
Expand
View Pricing