Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iminodiacetic acid
Synonym(s):
Iminodiacetic acid
Linear Formula:
HN(CH
2
COOH)
2
CAS Number:
142-73-4
Molecular Weight:
133.10
EC Number:
205-555-4
Products
Product Category
amino acids (2)
Brand
Sigma-Aldrich (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Purum (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
220000
Iminodiacetic acid,
98%, for peptide synthesis
Expand
View Pricing
8.14165
Iminodiacetic acid,
≥96.0% (acidimetric), for peptide synthesis
Expand
View Pricing
56781
Iminodiacetic acid,
≥98.0% (T), for peptide synthesis
Expand
View Pricing