Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hydroxyethyl-cellulose
Synonym(s):
Hydroxyethylcellulose
CAS Number:
9004-62-0
Products
Product Category
carbohydrates (2)
Brand
Sigma-Aldrich (3)
Color
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
09368
Hydroxyethyl-cellulose,
Expand
View Pricing
54290
Hydroxyethyl-cellulose,
viscosity 90-160 cP, 5 % in H
2
O(25 °C)
Expand
View Pricing
8.22068
Hydroxyethyl-cellulose,
for synthesis
Expand
View Pricing
Y0001951
Hydroxyethyl-cellulose,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing