Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hippuric acid
Synonym(s):
N
-Benzoylglycine, Benzoylaminoacetic acid, Hippuric acid
Linear Formula:
C
6
H
5
CONHCH
2
COOH
CAS Number:
495-69-2
Molecular Weight:
179.17
EC Number:
207-806-3
Products
Brand
Sigma-Aldrich (2)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
112003
Hippuric acid,
98%
Expand
View Pricing
8.20649
Hippuric acid,
for synthesis
Expand
View Pricing
68069
Hippuric acid,
analytical standard
Expand
View Pricing