Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hamilton
®
PRP-C18 (5 µm) HPLC Columns
Products
Product Category
columns (3)
Manufacturer
Separation Technique
ID (mm)
Length (mm)
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
50658-U
Hamilton
®
PRP-C18 (5 µm) HPLC Columns,
L × I.D. 50 mm × 2.1 mm, stainless steel
Expand
View Pricing
50672-U
Hamilton
®
PRP-C18 (5 µm) HPLC Columns,
L × I.D. 250 mm × 2.1 mm, PEEK
Expand
View Pricing
50662-U
Hamilton
®
PRP-C18 (5 µm) HPLC Columns,
L × I.D. 50 mm × 2.1 mm, PEEK
Expand
View Pricing