Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Greiner multiwell plate sealers
Products
Product Category
sealing films and foils (3)
Color
Manufacturer
Length (cm)
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
A5596
Greiner multiwell plate sealers,
EASYseal
™
sealing film, transparent, standard adhesive foil on carrier paper
Expand
View Pricing
Z617601
Greiner multiwell plate sealers,
SILVERseal
™
sealing film, adhesive aluminium foil on carrier paper, piercable, silver
Expand
View Pricing
Z617571
Greiner multiwell plate sealers,
VIEWseal
™
sealing film, pressure sensitive adhesive, transparent, double-perforated sides
Expand
View Pricing