Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Graphene
Linear Formula:
SiC
x
H
y
O
z
Products
Brand
Sigma-Aldrich (1)
Physical Form
Chemical Composition
Quality Segment
Functional Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
900561
Graphene,
powder, electrical conductivity >10
3
S/m, avg. no. of layers, < 3
Expand
View Pricing
900551
Graphene,
amine functionalized, avg. no. of layers, 1 ‑ 5
Expand
View Pricing
900552
Graphene,
silane modified, avg. no. of layers, 1 ‑ 5
Expand
View Pricing