Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gold-nickel alloy, Au82Ni18
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
GF70853418
Gold-nickel alloy, Au82Ni18,
foil, 50 x 50mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF31167514
Gold-nickel alloy, Au82Ni18,
0.1m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF26167305
Gold-nickel alloy, Au82Ni18,
0.1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF22609335
Gold-nickel alloy, Au82Ni18,
0.05m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF21953673
Gold-nickel alloy, Au82Ni18,
2m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF52245738
Gold-nickel alloy, Au82Ni18,
foil, 25 x 25mm, 0.25mm thickness, as rolled
Expand
View Pricing
GF62028659
Gold-nickel alloy, Au82Ni18,
foil, 50 x 50mm, 0.05mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF19853174
Gold-nickel alloy, Au82Ni18,
1m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF64158824
Gold-nickel alloy, Au82Ni18,
0.5m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF75416137
Gold-nickel alloy, Au82Ni18,
1m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF03195042
Gold-nickel alloy, Au82Ni18,
5m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF06208208
Gold-nickel alloy, Au82Ni18,
1m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF01649662
Gold-nickel alloy, Au82Ni18,
0.5m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF00111957
Gold-nickel alloy, Au82Ni18,
foil, 25 x 25mm, 0.05mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF70913279
Gold-nickel alloy, Au82Ni18,
0.2m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF23712498
Gold-nickel alloy, Au82Ni18,
0.2m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF43283752
Gold-nickel alloy, Au82Ni18,
0.5m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF99380874
Gold-nickel alloy, Au82Ni18,
10m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF76025561
Gold-nickel alloy, Au82Ni18,
2m wire, 0.025mm diameter, annealed
Expand
View Pricing