
Glycyrrhizic acid

Synonym(s):
3-O-(2-O-β-D-Glucopyranuronosyl-α-D-glucopyranuronosyl)-18β-glycyrrhetinic acid, Glycyrrhetinic acid glycoside, Glycyrrhizinic acid, Glycyrrhizic acid
Empirical Formula (Hill Notation):
C42H62O16
CAS Number:
Molecular Weight:
822.93
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2