
Glycyl-L-tyrosine dihydrate

Empirical Formula (Hill Notation):
C11H14N2O4 · 2H2O
CAS Number:
Molecular Weight:
274.27
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing