Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glass Packed GC Column (fits Agilent, Configuration "A")
Products
Product Category
columns (7)
Brand
Supelco (6)
Manufacturer
Agency
Column Type
ID (mm)
Length (m)
Compatibility
Column Material
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
13093-U
Glass Packed GC Column (fits Agilent, Configuration "A"),
phase none, matrix 80/100 Porapak Q support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
Expand
View Pricing
13087-U
Glass Packed GC Column (fits Agilent, Configuration "A"),
phase 3% OV-101, matrix 100/120 Chromosorb W HP support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
Expand
View Pricing
13088-U
Glass Packed GC Column (fits Agilent, Configuration "A"),
phase 10% Carbowax 20M, matrix 80/100 Chromosorb W HP support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
Expand
View Pricing
26012
Glass Packed GC Column (fits Agilent, Configuration "A"),
phase SP
®
-1000, 0.1%, matrix 80/100 Carbopack C support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
Expand
View Pricing
23110-U
Glass Packed GC Column (fits Agilent, Configuration "A"),
phase 4% Carbowax 20M, matrix 80/120 Carbopack B DA support, L × O.D. × I.D. 6.6 ft (2.0 m) × 1/4 in. × 2.0 mm, TightSpec
Expand
View Pricing
25936
Glass Packed GC Column (fits Agilent, Configuration "A"),
phase 4% Carbowax 20M, matrix 80/120 Carbopack B DA support, L × O.D. × I.D. 6.0 ft (1.8 m) × 1/4 in. × 2.0 mm
Expand
View Pricing
26021
Glass Packed GC Column (fits Agilent, Configuration "A"),
phase 4% Carbowax 20M + 0.8% KOH, matrix 60/80 Carbopack B support, L × O.D. × I.D. 6.6 ft (2.0 m) × 1/4 in. × 2.0 mm, TightSpec
Expand
View Pricing