Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Xplorer
®
Plus
Products
Product Category
pipettors (7)
Brand
Sigma-Aldrich (1)
Color
Manufacturer
Volume (µL)
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
EP4861000779
Eppendorf
®
Xplorer
®
Plus,
24-channel, adjustable, tip volume 1-20 μL, white
Expand
View Pricing
EP4861000778
Eppendorf
®
Xplorer
®
Plus,
16-channel, adjustable, tip volume 1-20 μL, white
Expand
View Pricing
EP4861000720
Eppendorf
®
Xplorer
®
Plus,
single-channel, adjustable, tip volume 10-200 μL, yellow
Expand
View Pricing
EP4861000710
Eppendorf
®
Xplorer
®
Plus,
single-channel, adjustable, tip volume 1-20 μL, light gray
Expand
View Pricing
EP4861000736
Eppendorf
®
Xplorer
®
Plus,
single-channel, adjustable, tip volume 0.125-2.5 mL, red
Expand
View Pricing
EP4861000792
Eppendorf
®
Xplorer
®
Plus,
16-channel, adjustable, tip volume 5-100 μL, light yellow
Expand
View Pricing
EP4861000793
Eppendorf
®
Xplorer
®
Plus,
24-channel, adjustable, tip volume 5-100 μL, light yellow
Expand
View Pricing