Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Xplorer
®
12-channel pipettor
Products
Product Category
pipettors (4)
Color
Manufacturer
Volume (µL)
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP4861000155
Eppendorf
®
Xplorer
®
12-channel pipettor,
volume 15-300 μL, multifunction rocker switch, orange, for pipette tips 300 ul
Expand
View Pricing
EP4861000171
Eppendorf
®
Xplorer
®
12-channel pipettor,
volume 50-1200 μL, multifunction rocker switch, green, for pipette tips 1250 ul
Expand
View Pricing
EP4861000139
Eppendorf
®
Xplorer
®
12-channel pipettor,
volume 5-100 μL, multifunction rocker switch, yellow, for pipette tips 200 ul
Expand
View Pricing
EP4861000112
Eppendorf
®
Xplorer
®
12-channel pipettor,
volume 0.5-10 μL, multifunction rocker switch, medium gray, for pipette tips 20 ul
Expand
View Pricing