Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Twin tec PCR plates, 384 wells
Products
Product Category
plates (14)
Manufacturer
Binding Type
Wells
Color
Plate Material
Package Size
Sterility
Technique
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
EP0030128508
Eppendorf
®
Twin tec PCR plates, 384 wells,
colorless, pkg of 25 ea
Expand
View Pricing
EP0030128516
Eppendorf
®
Twin tec PCR plates, 384 wells,
yellow plate, conical bottom yellow wells, pkg of 25 ea
Expand
View Pricing
EP0030128532
Eppendorf
®
Twin tec PCR plates, 384 wells,
blue plate, conical bottom blue wells, pkg of 25 ea
Expand
View Pricing
EP0030128966
Eppendorf
®
Twin tec PCR plates, 384 wells,
blue polypropylene plate, conical bottom colorless wells, pkg of 300 ea
Expand
View Pricing
EP0030128524
Eppendorf
®
Twin tec PCR plates, 384 wells,
green plate, conical bottom green wells, pkg of 25 ea
Expand
View Pricing
EP0030128818
Eppendorf
®
Twin tec PCR plates, 384 wells,
yellow plate, conical bottom black wells, pkg of 25 ea
Expand
View Pricing
EP0030128885
Eppendorf
®
Twin tec PCR plates, 384 wells,
orange plate, conical bottom colorless wells, pkg of 300 ea
Expand
View Pricing
EP0030128540
Eppendorf
®
Twin tec PCR plates, 384 wells,
red (wells colorless), pkg of 25 ea
Expand
View Pricing
EP0030128931
Eppendorf
®
Twin tec PCR plates, 384 wells,
colorless plate, conical bottom colorless wells, pkg of 300 ea
Expand
View Pricing
EP0030128893
Eppendorf
®
Twin tec PCR plates, 384 wells,
black plate, conical bottom colorless wells, pkg of 300 ea
Expand
View Pricing
EP0030128958
Eppendorf
®
Twin tec PCR plates, 384 wells,
green polypropylene plate, conical bottom colorless wells, pkg of 300 ea
Expand
View Pricing
EP0030128974
Eppendorf
®
Twin tec PCR plates, 384 wells,
red polypropylene plate, conical bottom colorless wells, pkg of 300 ea
Expand
View Pricing
EP0030128940
Eppendorf
®
Twin tec PCR plates, 384 wells,
yellow polypropylene plate, conical bottom colorless wells, pkg of 300 ea
Expand
View Pricing
EP0030128486
Eppendorf
®
Twin tec PCR plates, 384 wells,
with barcode, individual color (wells colorless), pkg of 25 ea
Expand
View Pricing