Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S.
®
G Singles, Biopur
®
Products
Product Category
pipette tips (3)
Color
Manufacturer
Binding Type
Volume (µL)
Color
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
EP0030010043
Eppendorf
®
epT.I.P.S.
®
G Singles, Biopur
®
,
volume (2-200 μL), L (53 mm), yellow Tray, pkg of 100 tips, individually wrapped
Expand
View Pricing
EP0030010060
Eppendorf
®
epT.I.P.S.
®
G Singles, Biopur
®
,
volume (50-1000 μL), L (71 mm), medium blue Tray, pkg of 100 tips, individually wrapped
Expand
View Pricing
EP0030010027
Eppendorf
®
epT.I.P.S.
®
G Singles, Biopur
®
,
volume (0.2-20 μL), L (40 mm), medium gray Tray, pkg of 100 tips, individually wrapped
Expand
View Pricing