Skip to Content
AM
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Epoxy embedding medium, accelerator
Synonym(s):
2,4,6-Tris(dimethylaminomethyl)phenol, DMP-30
Linear Formula:
[(CH
3
)
2
NCH
2
]
3
C
6
H
2
OH
CAS Number:
90-72-2
Molecular Weight:
265.39
EC Number:
202-013-9
Products
Product Category
mounting media (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
45348
Epoxy embedding medium, accelerator,
≥95% (NT)
Expand
View Pricing