![ECOSURF™ SA-9 Non-ionic surfactant](/deepweb/assets/sigmaaldrich/product/images/169/582/11c7bbf2-2cf3-4cfe-8a03-5385f1d2301a/640/11c7bbf2-2cf3-4cfe-8a03-5385f1d2301a.jpg)
ECOSURF™ SA-9
![ECOSURF™ SA-9 Non-ionic surfactant](/deepweb/assets/sigmaaldrich/product/images/169/582/11c7bbf2-2cf3-4cfe-8a03-5385f1d2301a/640/11c7bbf2-2cf3-4cfe-8a03-5385f1d2301a.jpg)
Synonym(s):
Alcohols ethoxylated propoxylated, Seed oil alcohol ethoxylate 9 EO
CAS Number:
Products
Product Category
Brand
Sustainability
Feature
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing